siRNA Details
Gene Symbol: Azgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012826.2 | NP_036958.2 | 2 | 160 | ||
XM_006249003.2 | XP_006249065.1 | 2 | 203 | |||
XM_006249004.2 | XP_006249066.1 | 2 | 202 | |||
GenBank mRNA | BC105822.1 | - | 2 | 160 | 7371444 | |
D21058.1 | - | 2 | 159 | |||
FQ210978.1 | - | 179 | ||||
FQ219509.1 | - | 2 | 151 | |||
X75309.1 | - | 2 | 141 |
