siRNA Details
Gene Symbol: Arrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012911.1 | NP_037043.1 | 2, 3 | 246 | ||
GenBank mRNA | BC087578.1 | - | 2, 3 | 118 | 7311618 | |
M91590.1 | - | 2, 3 | 246 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145429, NM_001271360, NM_001271358, NM_001271359, AK150389, and others.
Gene Ontology Categories:
