siRNA Details
Gene Symbol: LOC100364062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751952.3 | XP_003752000.1 | 1 | 1363 |
Gene Symbol: LOC100362738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772937.1 | XP_008771159.1 | 1 | 1403 |
Gene Symbol: Pkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053297.2 | NP_445749.1 | 10 | 1439 | ||
XM_006243187.2 | XP_006243249.1 | 10 | 1669 | |||
XM_006243188.2 | XP_006243250.1 | 10 | 1669 | |||
XM_006243190.2 | XP_006243252.1 | 11 | 1741 | |||
GenBank mRNA | AY724474.1 | - | 10 | 1436 | ||
BC061541.1 | - | 10 | 1314 | 6888187 | ||
FQ223721.1 | - | 10 | 1447 | |||
FQ224316.1 | - | 7 | 1097 | |||
FQ229350.1 | - | 1 | 39 | |||
X15800.1 | - | 10 | 1441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
