siRNA Details
Gene Symbol: Cpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013128.1 | NP_037260.1 | 3 | 690 | ||
GenBank mRNA | BC085762.1 | - | 3 | 689 | 7193151 | |
X51406.1 | - | 3 | 690 |

More Information
Other Species Targeted

Gene Ontology Categories:
