siRNA Details
Gene Symbol: Grn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145842.1 | NP_001139314.1 | 5 | 474 | ||
NM_017113.2 | NP_058809.2 | 5 | 474 | |||
XM_008767981.1 | XP_008766203.1 | 5 | 555 | |||
GenBank mRNA | BC072469.1 | - | 5 | 468 | 7098196 | |
CK365572.1 | - | 5 | 467 | |||
M97750.1 | - | 4 | 432 | |||
X62322.1 | - | 5 | 448 |
