siRNA Details
Gene Symbol: Ptpmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105726.1 | NP_001099196.1 | 3 | 945 | ||
GenBank mRNA | FQ219112.1 | - | 3 | 585 |

More Information
Other Species Targeted

Gene Ontology Categories:
