siRNA Details
Gene Symbol: CACFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242369.1 | NP_001229298.1 | 3 | 312 | ||
NM_017586.3 | NP_060056.1 | 3 | 312 | |||
XM_011518181.2 | XP_011516483.1 | 3 | 1869 | |||
XM_011518182.1 | XP_011516484.1 | 3 | 365 | |||
XM_011518185.2 | XP_011516487.1 | 3 | 1869 | |||
XM_011518186.1 | XP_011516488.1 | 3 | 365 | |||
GenBank mRNA | AJ011373.1 | - | 3 | 268 | ||
AK074852.1 | - | 3 | 290 | |||
AK075548.1 | - | 3 | 280 | |||
AK296343.1 | - | 3 | 287 | |||
AK304524.1 | - | 4 | 270 | |||
AK311325.1 | - | 3 | 368 | |||
AY358452.1 | - | 3 | 260 | |||
BC030558.1 | - | 3 | 278 | 5223979 | ||
DB165724.1 | - | 3 | 312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
