siRNA Details
Gene Symbol: Add2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109880.1 | NP_001103350.1 | 7 | 816 | ||
NM_012491.2 | NP_036623.1 | 7 | 816 | |||
GenBank mRNA | M63894.1 | - | 7 | 816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
