siRNA Details
Gene Symbol: Calca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033955.1 | NP_001029127.1 | 3 | 249 | ||
NM_001033956.1 | NP_001029128.1 | 3 | 225 | |||
NM_017338.2 | NP_059034.1 | 3 | 225 | |||
XM_008759676.1 | XP_008757898.1 | 3 | 276 | |||
GenBank mRNA | BG663259.1 | - | 3 | 191 | ||
CB711853.1 | - | 3 | 217 | |||
CF978038.1 | - | 3 | 230 | |||
M11597.1 | - | 2 | 123 | |||
M26137.1 | - | 2 | 130 | |||
V01228.1 | - | 3 | 186 | |||
V01229.1 | - | 2 | 130 | |||
V01231.1 | - | 2 | 122 |

More Information
Other Species Targeted

Gene Ontology Categories:
