siRNA Details
Gene Symbol: S100g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012521.2 | NP_036653.1 | 2 | 102 | ||
XM_006256888.2 | XP_006256950.1 | 2 | 477 | |||
GenBank mRNA | BC059153.1 | - | 2 | 106 | 6917928 | |
J02954.1 | - | 2 | 103 |

More Information
Other Species Targeted

Gene Ontology Categories:
