siRNA Details
Gene Symbol: Crygd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033095.1 | NP_149086.1 | 3 | 325 | ||
XM_008767074.1 | XP_008765296.1 | 2 | 1196 | |||
GenBank mRNA | X57169.1 | - | 3 | 310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
