siRNA Details
Gene Symbol: Prl8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022846.2 | NP_074037.1 | 5 | 449 | ||
XM_006253906.1 | XP_006253968.1 | 5 | 509 | |||
GenBank mRNA | BC097316.1 | - | 5 | 450 | 7442457 | |
FQ219955.1 | - | 5 | 509 | |||
FQ220067.1 | - | 5 | 508 | |||
FQ220110.1 | - | 5 | 509 | |||
FQ222955.1 | - | 5 | 509 | |||
FQ228468.1 | - | 1 | 79 | |||
FQ229664.1 | - | 5 | 483 | |||
FQ229702.1 | - | 5 | 507 | |||
FQ229765.1 | - | 5 | 506 | |||
FQ229882.1 | - | 5 | 509 | |||
L06441.1 | - | 5 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
