siRNA Details
Gene Symbol: Edn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012548.2 | NP_036680.1 | 3 | 549 | ||
GenBank mRNA | AF122903.1 | - | 1 | 95 | ||
M64711.1 | - | 3 | 544 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
