siRNA Details
Gene Symbol: Glra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012568.2 | NP_036700.1 | 8 | 1568 | ||
XM_003752039.3 | XP_003752087.1 | 9 | 1192 | |||
XM_008773152.1 | XP_008771374.1 | 8 | 1561 | |||
XM_008773153.1 | XP_008771375.1 | 8 | 974 | |||
GenBank mRNA | AJ310837.1 | - | 8 | 1017 | ||
X61159.1 | - | 8 | 1568 |

More Information
Other Species Targeted

Gene Ontology Categories:
