siRNA Details
Gene Symbol: Hoxb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191649.1 | NP_001178578.1 | 2 | 492 | ||
XM_006247175.2 | XP_006247237.1 | 2 | 501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
