siRNA Details
Gene Symbol: Iapp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012586.2 | NP_036718.1 | 2 | 98 | ||
XM_006237614.2 | XP_006237676.1 | 3 | 627 | |||
GenBank mRNA | J04544.1 | - | 3 | 185 | ||
M25390.1 | - | 3 | 134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
