siRNA Details
Gene Symbol: Ibsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012587.2 | NP_036719.2 | 5, 6 | 327 | ||
GenBank mRNA | AB001383.1 | - | 4, 5 | 254 | ||
BC127506.1 | - | 5, 6 | 323 | 8369751 | ||
DQ213013.1 | - | 4, 5 | 254 | |||
DY314698.1 | - | 5, 6 | 327 | |||
J04215.1 | - | 5, 6 | 333 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
