siRNA Details
Gene Symbol: Idh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031510.1 | NP_113698.1 | 1 | 105 | ||
XM_006245049.2 | XP_006245111.1 | 2 | 163 | |||
XM_008767076.1 | XP_008765298.1 | 2 | 195 | |||
GenBank mRNA | FQ214242.1 | - | 2 | 186 | ||
L35317.1 | - | 1 | 105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
