siRNA Details
Gene Symbol: Igf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190162.1 | NP_001177091.1 | 4 | 1121 | ||
NM_001190163.1 | NP_001177092.1 | 4 | 1109 | |||
NM_031511.2 | NP_113699.2 | 4 | 2096 | |||
XM_006230664.2 | XP_006230726.1 | 4 | 1898 | |||
XM_006230665.2 | XP_006230727.1 | 4 | 1101 | |||
XM_008760074.1 | XP_008758296.1 | 4 | 1919 | |||
XM_008760075.1 | XP_008758297.1 | 5 | 1434 | |||
GenBank mRNA | X13101.1 | - | 4 | 1134 | ||
X14833.1 | - | 4 | 1410 | |||
X14834.1 | - | 4 | 2165 | |||
X16703.1 | - | 1 | 748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
