siRNA Details
Gene Symbol: Il6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012589.2 | NP_036721.1 | 3, 4 | 391 | ||
GenBank mRNA | M26744.1 | - | 3, 4 | 391 |

More Information
Other Species Targeted

Gene Ontology Categories:
