siRNA Details
Gene Symbol: Jund | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286966.1 | NP_001273895.1 | 1 | 1429 | ||
NM_138875.4 | NP_620230.1 | 1 | 1429 | |||
GenBank mRNA | BC062053.1 | - | 1 | 1429 | 5598945 |

More Information
Other Species Targeted

Gene Ontology Categories:
