siRNA Details
Gene Symbol: Lalba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012594.1 | NP_036726.1 | 2 | 224 | ||
XM_008765668.1 | XP_008763890.1 | 3 | 2813 | |||
GenBank mRNA | V01251.1 | - | 1 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
