siRNA Details
Gene Symbol: Mbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012599.2 | NP_036731.2 | 5 | 453 | ||
GenBank mRNA | BC088159.1 | - | 5 | 453 | 7326244 | |
FQ209936.1 | - | 5 | 469 | |||
FQ218582.1 | - | 5 | 481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
