siRNA Details
Gene Symbol: Mme | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012608.2 | NP_036740.1 | 22, 23 | 2289 | ||
XM_006232437.2 | XP_006232499.1 | 22, 23 | 2325 | |||
GenBank mRNA | BC085753.1 | - | 22, 23 | 2289 | 7110842 | |
M15944.1 | - | 21, 22 | 2239 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
