siRNA Details
Gene Symbol: Nppa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012612.2 | NP_036744.1 | 2 | 227 | ||
GenBank mRNA | BC158590.1 | - | 2 | 217 | 9036379 | |
CB724799.1 | - | 2 | 354 | |||
M15868.1 | - | 1 | 41 | |||
X00665.1 | - | 2 | 219 | |||
X01118.1 | - | 2 | 208 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
