siRNA Details
Gene Symbol: Nppa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012612.2 | NP_036744.1 | 3 | 678 | ||
GenBank mRNA | BC158590.1 | - | 3 | 668 | 9036379 | |
M15868.1 | - | 2 | 492 | |||
M27498.1 | - | 3 | 666 | |||
X00665.1 | - | 3 | 669 | |||
X01118.1 | - | 3 | 658 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
