siRNA Details
Gene Symbol: Npr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012613.1 | NP_036745.1 | 5 | 1546 | ||
XM_006232591.2 | XP_006232653.1 | 6 | 2047 | |||
GenBank mRNA | BC128742.1 | - | 5 | 1532 | 7458324 | |
X14773.1 | - | 5 | 1403 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008727, BC110659, J05504, L31932.
Gene Ontology Categories:
