siRNA Details
Gene Symbol: Npr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012613.1 | NP_036745.1 | 16 | 2822 | ||
XM_006232591.2 | XP_006232653.1 | 17 | 3323 | |||
GenBank mRNA | BC128742.1 | - | 16 | 2808 | 7458324 | |
M74535.1 | - | 17 | 2771 | |||
X14773.1 | - | 16 | 2679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
