siRNA Details
Gene Symbol: Npr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012613.1 | NP_036745.1 | 10 | 2107 | ||
XM_006232591.2 | XP_006232653.1 | 11 | 2608 | |||
GenBank mRNA | BC128742.1 | - | 10 | 2093 | 7458324 | |
M74535.1 | - | 11 | 2056 | |||
X14773.1 | - | 10 | 1964 |

More Information
Other Species Targeted

Gene Ontology Categories:
