siRNA Details
Gene Symbol: Prl6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022176.2 | NP_071512.2 | 4 | 784 | ||
GenBank mRNA | BC158563.1 | - | 4 | 784 | 7443132 | |
FQ221549.1 | - | 4 | 801 | |||
FQ230053.1 | - | 4 | 802 | |||
FQ230138.1 | - | 11 | ||||
M31155.1 | - | 4 | 743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
