siRNA Details
Gene Symbol: Ppp2ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017039.2 | NP_058735.1 | 3 | 609 | ||
GenBank mRNA | BC070914.1 | - | 3 | 618 | 7104697 | |
BC072531.1 | - | 3 | 609 | 7096821 | ||
FQ228785.1 | - | 2 | 173 | |||
M33114.1 | - | 3 | 525 | |||
X14159.1 | - | 3 | 525 | |||
X16043.1 | - | 3 | 584 |
Gene Symbol: LOC103694903 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773894.1 | XP_008772116.1 | 1104 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
