siRNA Details
Gene Symbol: Ppp3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017042.2 | NP_058738.1 | 8 | 1035 | ||
GenBank mRNA | D90036.1 | - | 8 | 1035 | ||
FQ214219.1 | - | 84 | ||||
M31809.1 | - | 8 | 1035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
