siRNA Details
Gene Symbol: Ppy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012626.2 | NP_036758.1 | 4 | 434 | ||
GenBank mRNA | BP484139.1 | - | 4 | 417 | ||
M13588.1 | - | 4 | 434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
