siRNA Details
Gene Symbol: Prkar2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030020.1 | NP_001025191.1 | 8 | 861 | ||
GenBank mRNA | BC099223.1 | - | 7 | 604 | 7388087 | |
M12492.1 | - | 8 | 855 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
