siRNA Details
Gene Symbol: Prkcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012628.1 | NP_036760.1 | 6 | 840 | ||
XM_006228014.2 | XP_006228076.1 | 6 | 908 | |||
GenBank mRNA | BC089226.1 | - | 6 | 874 | 7319243 | |
M13707.1 | - | 6 | 840 | |||
X07287.1 | - | 6 | 763 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
