siRNA Details
Gene Symbol: Rbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012640.2 | NP_036772.2 | 2 | 182 | ||
XM_006243607.1 | XP_006243669.1 | 3 | 362 | |||
GenBank mRNA | M13949.1 | - | 2 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
