siRNA Details
Gene Symbol: Reg1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012641.1 | NP_036773.1 | 5 | 459 | ||
GenBank mRNA | FQ232485.1 | - | 5 | 451 | ||
M18962.1 | - | 4 | 435 | |||
M62930.1 | - | 5 | 459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
