siRNA Details
Gene Symbol: Shbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012650.1 | NP_036782.1 | 1, 2 | 140 | ||
XM_006246586.2 | XP_006246648.1 | 2, 3 | 889 | |||
GenBank mRNA | BC097336.1 | - | 1, 2 | 128 | 7453185 | |
M15034.1 | - | 1, 2 | 140 | |||
M31179.1 | - | 1, 2 | 92 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
