siRNA Details
Gene Symbol: Sod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017051.2 | NP_058747.1 | 5 | 705 | ||
GenBank mRNA | BC070913.1 | - | 5 | 705 | 7104698 | |
Y00497.1 | - | 5 | 633 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013671, CT010273, AK012354, AK167521, AK002428, and others.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
