siRNA Details
Gene Symbol: Serpina3n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031531.1 | NP_113719.1 | 2 | 684 | ||
GenBank mRNA | BC078796.1 | - | 2 | 636 | 7129287 | |
D00753.1 | - | 2 | 669 | |||
FQ209856.1 | - | 688 | ||||
HM448398.1 | - | 580 | ||||
X16359.1 | - | 2 | 684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
