siRNA Details
Gene Symbol: Tac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001124768.1 | NP_001118240.1 | 3 | 266 | ||
NM_001124769.1 | NP_001118241.1 | 3 | 266 | |||
NM_001124770.1 | NP_001118242.1 | 3 | 266 | |||
NM_012666.2 | NP_036798.1 | 3 | 266 | |||
XM_008762692.1 | XP_008760914.1 | 3 | 407 | |||
XM_008762693.1 | XP_008760915.1 | 3 | 404 | |||
XM_008762694.1 | XP_008760916.1 | 3 | 410 | |||
XM_008762695.1 | XP_008760917.1 | 3 | 408 | |||
GenBank mRNA | M14312.1 | - | 2 | 178 | ||
M15191.1 | - | 3 | 268 | |||
M34183.1 | - | 3 | 268 | |||
M34184.1 | - | 3 | 268 | |||
X56306.1 | - | 2 | 173 |

More Information
Other Species Targeted

Gene Ontology Categories:
