siRNA Details
Gene Symbol: Tgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012671.2 | NP_036803.1 | 6 | 1176 | ||
XM_008763050.1 | XP_008761272.1 | 6 | 2070 | |||
GenBank mRNA | M31076.1 | - | 6 | 1177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
