siRNA Details
Gene Symbol: Thy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012673.2 | NP_036805.1 | 3 | 210 | ||
GenBank mRNA | CB797376.1 | - | 3 | 210 | ||
M10666.1 | - | 1 | 60 | |||
M18002.1 | - | 1 | 60 | |||
X03150.1 | - | 3 | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
