siRNA Details
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012678.2 | NP_036810.1 | 7 | 654 | ||
GenBank mRNA | FQ225054.1 | - | 665 | |||
FQ225082.1 | - | 665 | ||||
J02780.1 | - | 7 | 657 |

More Information
Other Species Targeted

Gene Ontology Categories:
