siRNA Details
Gene Symbol: Clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053021.2 | NP_444180.2 | 4 | 464 | ||
XM_006252094.2 | XP_006252156.1 | 3 | 342 | |||
GenBank mRNA | BC061534.1 | - | 3 | 243 | 6886725 | |
FQ209467.1 | - | 3 | 240 | |||
FQ209498.1 | - | 3 | 241 | |||
FQ209623.1 | - | 3 | 239 | |||
FQ211071.1 | - | 3 | 239 | |||
FQ213433.1 | - | 3 | 238 | |||
FQ214098.1 | - | 3 | 284 | |||
FQ218237.1 | - | 3 | 239 | |||
FQ218812.1 | - | 3 | 239 | |||
FQ219030.1 | - | 3 | 239 | |||
M16975.1 | - | 4 | 464 | |||
M64723.1 | - | 3 | 229 | |||
X13231.1 | - | 3 | 228 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
