siRNA Details
Gene Symbol: Vcsa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012684.4 | NP_036816.4 | 3 | 226 | ||
GenBank mRNA | A07543.1 | - | 3 | 233 | ||
CB769246.1 | - | 3 | 238 | |||
M63112.1 | - | 3 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
