siRNA Details
Gene Symbol: F9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031540.1 | NP_113728.1 | 8 | 1326 | ||
GenBank mRNA | M26247.1 | - | 3 | 810 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007979, AK149372, BC132393, M26236, BC125617, and others.
Gene Ontology Categories:
