siRNA Details
Gene Symbol: Gcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012707.2 | NP_036839.2 | 3, 4 | 317 | ||
GenBank mRNA | BP479145.1 | - | 3, 4 | 317 | ||
BP480555.1 | - | 3, 4 | 292 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
