siRNA Details
Gene Symbol: RT1-A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008827.1 | NP_001008827.1 | 8 | 1458 | ||
XM_006256099.2 | XP_006256161.1 | 7 | 1441 | |||
GenBank mRNA | FQ228888.1 | - | 8 | 1505 | ||
FQ231496.1 | - | 6 | 1112 | |||
KC222922.1 | - | 7 | 1363 | |||
U50448.1 | - | 8 | 1449 |
Gene Symbol: RT1-A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008830.1 | NP_001008830.1 | 9 | 1460 |
Gene Symbol: RT1-EC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | L26224.1 | - | 8 | 1470 | ||
L40363.1 | - | 9 | 1451 | |||
M24023.1 | - | 7 | 1240 | |||
U38970.1 | - | 8 | 1454 | |||
U50447.1 | - | 10 | 1449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
