siRNA Details
Gene Symbol: RT1-M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001717.1 | NP_001001717.1 | 4 | 764 | ||
XM_008772690.1 | XP_008770912.1 | 3 | 872 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
